Order Entry
Austria
ContactUsLinkComponent
7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
Catalog # AATB553
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
7-Hydroxycoumarin-4-acetic acid succinimidyl ester ≥90%
Catalog # AATB553
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :317.25 g/mol