Order Entry
Austria
ContactUsLinkComponent
Fmoc-4,5-dehydro-l-leucine ≥95%
Fmoc-4,5-dehydro-l-leucine ≥95%
Catalog # AATB5300
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Fmoc-4,5-dehydro-l-leucine ≥95%
Catalog # AATB5300
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :351.402 g/mol
ADR:5,II
 :GHS07
CAS Number:87720-55-6
Formula:C₂₁H₂₁NO₄
MDL Number:MFCD00237651