Order Entry
Austria
ContactUsLinkComponent
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride for oligonucleotide labeling
Irritant
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride for oligonucleotide labeling
Catalog # AATB4021
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
3,6-Dichlorobenzene-1,2,4-tricarboxylic anhydride for oligonucleotide labeling
Catalog # AATB4021
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :261.0152 g/mol
ADR:5,II
 :GHS07
CAS Number:81742-10-1
Formula:C₉H₂Cl₂O₅
MDL Number:MFCD28962732