Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Biotin azide
Biotin azide biodetection
Biotin azide biodetection
Biotin azide biodetection
Catalog # AATB3020
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Biotin azide biodetection
Catalog # AATB3020
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
ADR:
5,II
:
326.42 g/mol
:
GHS07
Product Details