Order Entry
Austria
ContactUsLinkComponent
Phenol Red, sodium salt
Irritant
Phenol Red, sodium salt
Catalog # AATB2460
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Phenol Red, sodium salt
Catalog # AATB2460
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :376.36 g/mol
ADR:5,II
 :GHS07
CAS Number:34487-61-6
Formula:C₁₉H₁₃O₅SNa