Order Entry
Austria
ContactUsLinkComponent
 
 
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%
  AATB22220
 :  
undefined
 
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%
  AATB22220
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

 

 

 :
 :514.95 g/mol
 :5,II
 :GHS07
 :115532-52-0
 :C₂₆H₂₇ClN₂O₇
 :MFCD00467973