Order Entry
Austria
ContactUsLinkComponent
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) mixture of isomers ≥90%
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) mixture of isomers ≥90%
Catalog # AATB22026
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester (CDCFDA) mixture of isomers ≥90%
Catalog # AATB22026
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :626.35 g/mol
CAS Number:147265-60-9
Formula:C₂₉H₁₇Cl₂NO₁₁