0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
6-NED azide
6-NED azide fluorescent probe
6-NED azide fluorescent probe
AATB217
:
AAT BIOQUEST
:
6-NED azide fluorescent probe
AATB217
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
5,II
:
615.78 g/mol
:
GHS07