
BCECF AM ester, Ultra Pure Grade calcium indicator
Artikelnr. AATB21203
Supplier: AAT BIOQUEST
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1.000 µG
- Seq:1442
Specifications
About this item
:
:808.69 g/mol
ADR:5,II
:GHS07
CAS Number:117464-70-7
Formula:C₄₂H₄₀O₂₁
MDL Number:MFCD00036969