Order Entry
Austria
ContactUsLinkComponent
Indo-1 pentapotassium salt ≥95%
Irritant
Indo-1 pentapotassium salt ≥95%
Catalog # AATB21040
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Indo-1 pentapotassium salt ≥95%
Catalog # AATB21040
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :840.05 g/mol
ADR:5,II
 :GHS07
CAS Number:132319-56-3
Formula:C₃₂H₂₆K₅N₃O₁₂