Order Entry
Austria
ContactUsLinkComponent
Calbryte™-520L potassium salt
Calbryte™-520L potassium salt
Catalog # AATB20642
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Calbryte™-520L potassium salt
Catalog # AATB20642
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 µG
  • Seq:
    1418

Specifications

About this item

 :
 :794.88 g/mol
ADR:5,II
 :GHS07