Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Supplier:   SI Analytics
Electrode Cables
Supplier:  SI Analytics
Certificates
Electrode Cables
Electrode Cables
19 Options Available Below
Product Details & Documents
Electrode cables with a range of connectors to fit most pH meters.
Product Family Options
Coaxial cables for pH electrodes 1 metre ( L type)
Product Information
  • Length
    Availability
    Price
  • 662-6118
    Without connector (free end) for custom installation
    Without connector (free end) for custom installation
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,40 €
    42,40 €
    1 * 1 ST
       42,40 €
  • 662-0102
    DIN connector for Schott, WTW pH meters
    DIN connector for Schott, WTW pH meters
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,00 €
    67,00 €
    1 * 1 ST
       67,00 €
  • 662-6193
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 67,00 €
    67,00 €
    1 * 1 ST
       67,00 €
  • 662-6117
    R-connector for Metrohm (f-plug) meters
    R-connector for Metrohm (f-plug) meters
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 82,00 €
    82,00 €
    1 * 1 ST
       82,00 €
Single core cables for Reference electrodes ( B type)
Product Information
  • Length
    Availability
    Price
  • 662-6124
    Electrode cable with 2 mm pole connector
    Electrode cable with 2 mm pole connector
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,80 €
    39,80 €
    1 * 1 ST
       39,80 €
  • 662-6119
    Electrode cable with 4 mm banana connector
    Electrode cable with 4 mm banana connector
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,70 €
    38,70 €
    1 * 1 ST
       38,70 €
  • SCOI285121854
    Without connector (free end) for custom installation of reference electrode plug
    Without connector (free end) for custom installation of reference electrode plug
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 44,80 €
    44,80 €
    1 * 1 ST
       44,80 €
Coaxial cables for pH electrodes ( K type)
Product Information
  • Length
    Availability
    Price
  • SCOI285120011
    Without connector (free end) for custom installation
    Without connector (free end) for custom installation
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,70 €
    49,70 €
    1 * 1 ST
       49,70 €
  • SCOI285120028
    Without connector (free end) for custom installation
    Without connector (free end) for custom installation
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 56,00 €
    56,00 €
    1 * 1 ST
       56,00 €
  • SCOI285120036
    Without connector (free end) for custom installation
    Without connector (free end) for custom installation
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 64,00 €
    64,00 €
    1 * 1 ST
       64,00 €
  • SCOI285120052
    Without connector (free end) for custom installation
    Without connector (free end) for custom installation
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,00 €
    76,00 €
    1 * 1 ST
       76,00 €
  • 662-0061
    DIN connector for Schott, WTW pH meters
    DIN connector for Schott, WTW pH meters
    1 m
    Log in for availability
    Log in for availability
    1 * 1 m
    1 * 1 m - 70,00 €
    70,00 €
    1 * 1 m
       70,00 €
  • SCOI285120225
    DIN connector for Schott, WTW pH meters
    DIN connector for Schott, WTW pH meters
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,00 €
    76,00 €
    1 * 1 ST
       76,00 €
  • SCOI285120233
    DIN connector for Schott, WTW pH meters
    DIN connector for Schott, WTW pH meters
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,00 €
    85,00 €
    1 * 1 ST
       85,00 €
  • SCOI285120258
    DIN connector for Schott, WTW pH meters
    DIN connector for Schott, WTW pH meters
    5 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 99,00 €
    99,00 €
    1 * 1 ST
       99,00 €
  • 662-0062
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 70,00 €
    70,00 €
    1 * 1 ST
       70,00 €
  • SCOI285121221
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    BNC connector for pH meters with BNC for example Hach, Orion, Mettler
    2 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 76,00 €
    76,00 €
    1 * 1 ST
       76,00 €
  • SCOI285121616
    S-connector, socket for cable extension
    S-connector, socket for cable extension
    1 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 88,00 €
    88,00 €
    1 * 1 ST
       88,00 €
  • SCOI285121632
    S-connector, socket for cable extension
    S-connector, socket for cable extension
    3 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 101,00 €
    101,00 €
    1 * 1 ST
       101,00 €