Order Entry
Austria
Orders LinkContactUsLinkComponent
Formaldehyde solution 4%, ReadiUse™ fixative reagent

Formaldehyde solution 4%, ReadiUse™ fixative reagent

Plastic bottle, 50 mL
Catalog # AATB20010
Irritant
Supplier:   AAT Bioquest
Formaldehyde solution 4%, ReadiUse™ fixative reagent
Catalog # AATB20010
Supplier:  AAT Bioquest
CAS Number:  50-00-0
Formaldehyde solution 4%, ReadiUse™ fixative reagent
Formaldehyde solution 4%, ReadiUse™ fixative reagent

Plastic bottle, 50 mL

CAS Number:  50-00-0
Price
122,00 €
Log in for availability
1 * 50 mL
Product Details & Documents

All our ReadiUse™ reagents require minimal hands-on time.

  • Non-reactive
  • Long-term stability
  • Minimal hands-on time
  • Ready to use

Caution: For research use only.

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.