Order Entry
Austria
ContactUsLinkComponent
DiYO™-3 5 mM in DMSO ≥90%
DiYO™-3 5 mM in DMSO ≥90%
Catalog # AATB17581
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
DiYO™-3 5 mM in DMSO ≥90%
Catalog # AATB17581
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    0,2 mL
  • Seq:
    2020

Specifications

About this item

 :
 :1322.73 g/mol
ADR:5,II
 :GHS07
CAS Number:156312-20-8
Formula:C₅₃H₅₈I₄N₆O₂