Order Entry
Austria
ContactUsLinkComponent
Propidium iodide 10 mM in aqueous solution ≥95%
Propidium iodide 10 mM in aqueous solution ≥95%
Catalog # AATB17517
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Propidium iodide 10 mM in aqueous solution ≥95%
Catalog # AATB17517
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :668.4 g/mol
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921