0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Austria
ContactUsLinkComponent
DAX-J2™
DAX-J2™ IR
DAX-J2™ IR
DAX-J2™ IR
Catalog # AATB16302
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
DAX-J2™ IR
Catalog # AATB16302
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details