Order Entry
Austria
ContactUsLinkComponent
ROS Brite™ APF Optimized for Detecting Reactive Oxygen Species (ROS)
ROS Brite™ APF Optimized for Detecting Reactive Oxygen Species (ROS)
Catalog # AATB16050
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
ROS Brite™ APF Optimized for Detecting Reactive Oxygen Species (ROS)
Catalog # AATB16050
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :GHS07