Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
ROS Brite™
ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
Irritant
ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
Catalog # AATB16002
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
Irritant
ROS Brite™ 670 Optimized for Detecting Reactive Oxygen Species (ROS)
Catalog # AATB16002
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details