Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
GMP-Fluorescein
GMP-Fluorescein conjugate for calibration
GMP-Fluorescein conjugate for calibration
GMP-Fluorescein conjugate for calibration
Catalog # AATB13611
Supplier:
AAT BIOQUEST
CAS Number:
Expand image
GMP-Fluorescein conjugate for calibration
Catalog # AATB13611
Supplier:
AAT BIOQUEST
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 Nmo
Seq:
9040
Specifications
About this item
Product Details
:
:
800 g/mol
ADR:
5,II
:
GHS07
Product Details