Order Entry
Austria
Orders LinkContactUsLinkComponent
Z-DEVD-FMK

Z-DEVD-FMK

Plastic bottle, 1 mg
Catalog # AATB13303
Irritant
Supplier:   AAT Bioquest
Z-DEVD-FMK
Catalog # AATB13303
Supplier:  AAT Bioquest
CAS Number:  210344-95-9
Z-DEVD-FMK
Z-DEVD-FMK

Plastic bottle, 1 mg

CAS Number:  210344-95-9
Price
147,00 €
Log in for availability
1 * 1 mg
Product Details & Documents

Z-DEVD-FMK is a specific, irreversible Caspase-3 inhibitor, and also shows potent inhibition on caspase-6, caspase-7, caspase-8, and caspase-10.

  • Multiple caspases inhibitor
  • Enhanced stability
  • Increased cell permeability
  • CAS Number:210344-95-9
  • Formula:C₃₀H₄₁FN₄O₁₂
  • MDL Number:MFCD23380217
  • Molecular Weight:668.66 g/mol
  • ADR:5,II
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg

Specifications