Order Entry
Austria
ContactUsLinkComponent
SPINeasy® PCR Purification and Gel Extraction Kit, MP Biomedicals
SPINeasy® PCR Purification and Gel Extraction Kit, MP Biomedicals
Catalog # 116538050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® PCR Purification and Gel Extraction Kit, MP Biomedicals
Catalog # 116538050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Enzymatic clean-up
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Description:
    SPINeasy® PCR purification and gel extraction kit, 50 tests
  • Storage temperature:
    Store at room temperature (15...30 °C), in a dry environment.

Specifications

About this item

SPINeasy PCR purification and gel extraction Kit is a silica-membrane spin-column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gel electrophoresis.

Upto 23 µg of DNA of molecular weight ranging from 100 bp to 20 kb can be purified through a quick and simple process. Purified DNA is immediately ready for routine molecular biology laboratory applications.

SPINeasy PCR purification and gel extraction kit is a silica-membrane spin-column kit that enables quick and convenient DNA clean up from various enzymatic reactions, such as PCR and restriction digestion, as well as isolation and purification of DNA fragments from agarose gel electrophoresis.

Certifications: Hazard Class 8
Delivery: Kit includes 50 ml Binding Buffer PG, 6 ml Wash Buffer PG, 10 ml Elution Buffer PG, 50× Column PG with collection tubes.