Demonstration thermometer
Demonstration thermometer
Catalog # 765-1079
Marketsource
Supplier: MOLLER THERM
Jump to:
Product Details & Documents
Milk glass scale on polished wooden board. Colored liquid filling. Measuring range -10 to +100 ° C, 0 to +80 ° R, +20 to +212 ° F.
Specifications
- Description:Demonstration thermometer
- Cat. No.:765-1079