Clips for conical standard ground joints
BRND55646
: Brand
- For NS:29/32
- Colour:Red
- Pk:1
POM. Will not scratch glass surfaces.
- Excellent resilience and stability
- Outstanding chemical resistance
- Operating temperature up to 150 °C
POM. Will not scratch glass surfaces.