Sprayer
Sprayer, complete
Catalog # 217-0695
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass sprayer.
Delivery information: Complete sprayer consists of 100 ml Erlenmeyer flask with socket NS 19/26, sprayer head with clamp and rubber bulb.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Description:Sprayer, complete
Accessories
Product Information
- Catalog #AvailabilityPrice
- Sprayer head, NS 19/26, without flask or rubber bulbSprayer head, NS 19/26, without flask or rubber bulb201-1434Log in for availability1 * 1 ST - 45,70 €
Product Information
- Catalog #AvailabilityPrice
- Rubber bulb