Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottle acc. to Winkler, uncalibrated, without numbering, 100 ml, 19/26
Catalog # 705-0570
Dissolved oxygen bottles
Catalog # 705-0570
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottle acc. to Winkler, uncalibrated, without numbering, 100 ml, 19/26
Price
13,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Bottle, approx. 100 ml capacity, made from clear soda lime glass with grounded glass stopper. Height with stopper approx. 115 to 120 mm. Accessories include a clamp that holds the stopper in place.
Specifications
  • Capacity:
    100 ml
  • Description:
    Oxygen bottle acc. to Winkler, uncalibrated, without numbering
  • NS stopper:
    19/26