Drop-Out Mix Complete without Yeast Nitrogen Base
Catalog # USBID9515-100G
Supplier: US Biological
CAS Number:
Specifications
- Pk:100 g
- Description:Drop-out mix complete without yeast nitrogen base (DO, dropout, powder)
Specifications
About this item
Drop-out mixes are used as a supplement to Drop-out Base for making synthetic, complete media formulations in accordance with methods in yeast genetics.
- Convenient powders
- Useful for one-hybrid and two-hybrid systems
All lots are quality control tested using the appropriate yeast strains and transformants.