Order Entry
Austria
ContactUsLinkComponent
 
SPINeasy® DNA Purification Kit, MP Biomedicals
Catalog # 116548050.
Supplier:  MP Biomedicals
CAS Number:  
undefined
SPINeasy® DNA Purification Kit, MP Biomedicals
Catalog # 116548050.
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Method / Format:
    Enzymatic clean-up
  • Sample type:
    DNA preps
  • Target molecule:
    DNA
  • Description:
    SPINeasy® DNA purification kit, 50 tests
  • Storage temperature:
    15...25 ℃

Specifications

About this item

Humic acids, heavy metals, heme are the most notorious PCR inhibitors. This could be due to non-optimised DNA purification procedures which often co-purify inhibitors and lead to false negative results on downstream applications.

  • Efficient clean-up of highly contaminated pre-purified DNA sample
  • Clean: Effective removal of inhibitory compound
  • Buffer P1 and P1HA suitable for sample with various degree of contamination

The SPINeasy® DNA Purification Kit is formulated to effectively remove contaminants on pre-purified DNA samples using the novel and proprietary humic acid removal technology. It can recover high quality DNA for all downstream applications.