ContactUsLinkComponent
VWR® BaseBond Micro Vials with Crimp Neck, ND11
VWR® BaseBond Micro Vials with Crimp Neck, ND11
Catalog # 548-2648A
Supplier:  VWR Collection
VWR® BaseBond Micro Vials with Crimp Neck, ND11
Catalog # 548-2648A
Supplier:  VWR Collection
Specifications
  • Ø×H:
    11,6×32,0 mm
  • Packed:
    100 per pack
  • Capacity:
    0,20 ml
  • Colour:
    Amber
  • Description:
    Crimp neck vial with integrated microinsert
  • Material:
    Glass
  • Pk:
    1.000
Specifications
About this item
  • 1st hydrol. class, writing patch, with small micro-insert
  • BaseBond