Order Entry
Austria
ContactUsLinkComponent
 
Sodium thiosulphate pentahydrate 0,005 mol/L, colourless, liquid
Catalog # CMPLZ15321110.1L
CAS Number:  
Sodium thiosulphate pentahydrate 0,005 mol/L, colourless, liquid
Catalog # CMPLZ15321110.1L
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Density:
    1 g/ml
  • pH:
    8 - 9
  • Soulubility:
    Soluble

Specification Test Results

About this item

 :
 :248.19 g/mol
Boiling Pt:100 °C
Melting Pt:48,3 °C
CAS Number:10102-17-7
Formula:Na₂S₂O₃.5H₂O
MDL Number:MFCD00149186