Specifications
- Weight:6,2 kg
- Temperature range:15 °C below ambient to 100 °C
- Description:ThermoMixer™ C, without thermoblock
- W×D×H:206×304×131 mm
- Electrical:120 V, 50/60 Hz
- Pk:1
Specifications
About this item
The Eppendorf® thermomixer™ C improves assay results by mixing and incubating samples at the same time.
Units allow heating, mixing, and cooling in all common vessel and plate formats from 5 μl to 50 ml, with high-speed mixing up to 3000 min⁻¹ (depending on the SmartBlock™ used) for fast mixing, even in 384-well plates. Excellent mixing performance is due to unique 2D Mix-Control technology. The optional ThermoTop lid reliably prevents condensate formation. Simple and intuitive operation is performed via pre-defined program keys and a clearly arranged menu. Units also offer versatile program functions, the ability to save up to twenty user-defined programs, and a two-second block exchange thanks to the SmartBlock™ quick release system. The ergonomic design and optimized features are engineered in accordance with the PhysioCare Concept.
The ThermoMixer™ C accepts quick and easy-to-exchange SmartBlocks™ for all common lab vessels, along with new and improved program functions and a high level of stability for every mixing frequency.