Order Entry
Austria
ContactUsLinkComponent
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide Phenyl-Hexyl Fused-Core®
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide Phenyl-Hexyl Fused-Core®
Supplier:  Merck
Certificates
Supelco® BIOshell™ HPLC Columns, 160 Å Peptide Phenyl-Hexyl Fused-Core®
Supplier:  Merck

Specifications

  • Catalog No:
  • 577543-U
  • 577539-U
  • 577540-U
  • 577541-U
  • 577537-U
  • 577542-U
  • 577538-U
  • 577535-U
  • 577536-U
  • 577533-U
  • 577534-U
  • 577545-U
  • 577525-U
  • 577526-U
  • 577523-U
  • 577524-U
  • 577550-U
  • 577548-U
  • 577547-U
  • 577546-U
  • 577531-U
  • 577532-U
  • 577529-U
  • 577530-U
  • 577527-U
  • 577528-U
  • 577549-U
  • 577544-U
  • Application:
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
  • Carbon load:
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • 4,7%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Max. pressure:
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • 600 bar
  • pH range:
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • 2 - 9
  • Surface area:
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • 90 m²/g
  • Temperature limit:
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • 90 °C
  • USP code:
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11
  • L11

Specifications

Guard Cartridge Holder

Product Information