SGE OptChem™ Double Taper Inlet Liners
The SGE inlet liner range aims to make it simple for all gas chromatographers to select the correct liner.
- Easy to choose - colour coded by geometry to simplify your selection
- Easy to use - contains O-rings so ready to go
- Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the head of the column
- Top taper aids in minimizing sample flashback
- Color coded yellow
Double taper liners are recommended for the following sample or injection modes: trace level analyses, splitless, low boiling point compounds, active compounds.
: Double taper available for Agilent, Thermo Scientific, and Varian/Bruker 1177 and 1078/1079 injectors.
CRS O-Rings
Frequently Bought Together