CRS ONE O-Rings for Inlet Liner Seals
The o-ring prevents gas leaks and maintains a uniform flow path through the inlet, where even small leaks can cause sample loss - affecting accuracy and reproducibility of results.
- Consistently forms a reliable seal, preventing inlet leaks and sample loss
- Pre-cleaned and conditioned to minimize potential active sites and system contamination
- Resistant to high temperatures and chemical exposure, ensuring optimal performance, durability and longevity
- Compatible with multiple instruments
Applications: GC and GC-MS analysis. Ideal for trace level analysis and applications where high levels of cleanliness and sensitivity are required.
FKM o-rings available for industry standard inlet liner sizes and various injectors. Maximum injector temperature of 300 °C.
: Available in packs of 10.
Frequently Bought Together