- Shoe size:44
- Colour:Black
- Pk:1 PAIR
Safety shoes made from leather with replaceable insole with fabric antibacterial lining and microfiber upper material.
- Steel toe cap
- Antibacterial lining
- Antibacterial insole
- Orthopedic insoles used
- Washable 30°
- HACCP compliant
Suitable for food industry. Slip resistance on ceramic, coated with SLS solution and on steel, coated with glycerol - slip resistance on ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol.
Colour: Black
Fastener type: Shoelaces
Weight of half a pair [kg] (for the size): 0,4550 (42)
Upper material: Microfiber
Lining material: Fabric
Lining material (features): Antibacterial lining
Insole: Fabric
Antistatic footwear - antistatic properties
Energy absorption in the heel area - energy absorbtion in the heel area
Water permeability and water absorption of the face material - permeability and water absorption of the upper material
Toe: Steel, strong, durable and impact-resistant with 200J energy, comfortable thanks to elastomer finishing, which makes the edges imperceptible with bent feet.
Protection class: S2 - 200 joule toecap protection. Closed seat region (fully enclosed heel). Anti-static properties. Energy absorption of seat region. Water penetration and water absorption resistance.