Cell culture dishes, glass bottom, CELLview™
Supplier: Greiner Bio-One
Certificates
About this item
PS with integrated glass bottom, sterile. CELLview™ glass bottom dishes combine the convenience of a standard 35 mm disposable plastic cell culture dish with the optical quality of glass, providing superior high resolution microscopic images of in vitro cultures. The CELLview™ dish is also available as a subdivided version with four individual compartments facilitating simultaneous multiplex analysis of different cell lines, various stimulations or diverse transfections, minimising the amount of required cells and reagents per individual assay.
- High transparent achromatic borosilicate glass base; hydrolytic class 1 (DIN-ISO 719)
- Maximal spectral transmission; no autofluorescence; no depolarisation of light
- Available with specific surface treatments increasing the attachment of adherent cells
- Broad range of multiple microscopic applications
Manufactured according to ISO 8255-1:1986
Certifications: Manufactured according to ISO 8255-1:1986
Specifications
- Catalog No:
- 391-0254
- 391-0253
- 391-0252
- 391-0255
- 391-0257
- 391-0256
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)