Order Entry
Austria
ContactUsLinkComponent
Mouse Anti-PEG IgM ELISA kit
Mouse Anti-PEG IgM ELISA kit
Catalog # ABNOKA6895.1KIT
Supplier:  Abnova
CAS Number:  
Mouse Anti-PEG IgM ELISA kit
Catalog # ABNOKA6895.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Anti-PEG IgM

Specifications

About this item

Mouse Anti-PEG IgM ELISA Kit can be used for detection of anti-PEG IgM in mouse serum, plasma and tissue fluids.