Order Entry
Austria
ContactUsLinkComponent
Rat Anti-PEG IgG ELISA kit
Rat Anti-PEG IgG ELISA kit
Catalog # ABNOKA6896.1KIT
Supplier:  Abnova
CAS Number:  
Rat Anti-PEG IgG ELISA kit
Catalog # ABNOKA6896.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Rat
  • Target protein:
    Anti-PEG IgG

Specifications

About this item

Rat Anti-PEG IgG ELISA Kit can be used for detection of anti-PEG IgG in rat serum, plasma and tissue fluids.