Rat Anti-PEG IgM ELISA kit
Catalog # ABNOKA6897.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Target protein:Anti-PEG IgM
Specifications
About this item
Rat Anti-PEG IgM ELISA Kit can be used for detection of anti-PEG IgM in rat serum, plasma and tissue fluids.