Order Entry
Austria
ContactUsLinkComponent
Monkey Anti-PEG IgM ELISA kit
Monkey Anti-PEG IgM ELISA kit
Catalog # ABNOKA6899.1KIT
Supplier:  Abnova
CAS Number:  
Monkey Anti-PEG IgM ELISA kit
Catalog # ABNOKA6899.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    Anti-PEG IgM

Specifications

About this item

Monkey Anti-PEG IgM ELISA kit can be used for detection of anti-PEG IgM in monkey serum, plasma and tissue fluids.