Order Entry
Austria
ContactUsLinkComponent
The IgA (Monkey) ELISA kit
The IgA (Monkey) ELISA kit
Catalog # ABNOKA6747.1KIT
Supplier:  Abnova
CAS Number:  
The IgA (Monkey) ELISA kit
Catalog # ABNOKA6747.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Monkey
  • Target protein:
    IgA

Specifications

About this item

The IgA (Monkey) ELISA Kit is used for determination of IgA in old world monkey serum or plasma.