IgM (Cow) ELISA kit
ABNOKA6763.1KIT
: Abnova
:
- VE:1 KIT
- Reaktivität der primären Antikörper:Bovine
- Target protein:IgM
- Testprinzip:Quantitative
IgM (Cow) ELISA Kit is for the quantitative measurement of cow IgM in serum or plasma.
IgM (Cow) ELISA Kit is for the quantitative measurement of cow IgM in serum or plasma.