Order Entry
Austria
ContactUsLinkComponent
 
Herpes Simplex Virus I IgG ELISA kit
Catalog # ABNOKA6733.1KIT
Supplier:  Abnova
CAS Number:  
Herpes Simplex Virus I IgG ELISA kit
Catalog # ABNOKA6733.1KIT
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    HSV1
  • Target protein:
    IgG
  • Assay Principle:
    Qualitative

Specifications

About this item

Herpes Simplex Virus I IgG ELISA Kit is a qualitative determination of IgG class antibodies against Herpes simplex Virus 1 (HSV 1) in human serum or plasma.