Cytomegalovirus IgM ELISA kit
Catalog # ABNOKA6725.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Cytomegalovirus
- Target protein:IgM
- Assay Principle:Qualitative
Specifications
About this item
Cytomegalovirus IgM ELISA Kit is a qualitative determination of IgM class antibodies against Cytomegalovirus (CMV) in human serum or plasma.