Candida albicans IgG ELISA kit
Catalog # ABNOKA6722.1KIT
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Candida albicans
- Target protein:IgG
- Assay Principle:Qualitative
Specifications
About this item
Candida albicans IgG ELISA Kit is a qualitative determination of IgG class antibodies against Candida albicans in human serum or plasma.