Order Entry
Austria
ContactUsLinkComponent
Recombinant Human HGF
Recombinant Human HGF
Catalog # ABNOP8759.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Human HGF
Catalog # ABNOP8759.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein/peptide name:
    HGF

Specifications

About this item