Order Entry
Austria
ContactUsLinkComponent
Recombinant Human IGFBP3 (from E. coli), His-tag
Recombinant Human IGFBP3 (from E. coli), His-tag
  ABNOP8193.25UG
 :  Abnova
 :  
Recombinant Human IGFBP3 (from E. coli), His-tag
  ABNOP8193.25UG
 :  Abnova
 :  

 

  • Pk:
    25 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Tag sequence:
    His-tag
  • Gene ID:
    P17936
  • Protein/peptide name:
    IGFBP3
  • Sequence:
    28a.a.-291a.a.