Order Entry
Austria
ContactUsLinkComponent
Recombinant Mouse IL1A (from E. coli), His-tag
Recombinant Mouse IL1A (from E. coli), His-tag
  ABNOP8209.20UG
 :  Abnova
 :  
Recombinant Mouse IL1A (from E. coli), His-tag
  ABNOP8209.20UG
 :  Abnova
 :  

 

  • Pk:
    20 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Tag sequence:
    His-tag
  • Gene ID:
    P01582
  • Protein/peptide name:
    IL1A
  • Sequence:
    115a.a.-270a.a.