Recombinant Mouse IL1B (from E. coli), His-tag
Catalog # ABNOP8222.5UG
Supplier: Abnova
CAS Number:
Specifications
- Pk:5 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Tag sequence:His-tag
- Gene ID:P10749
- Protein/peptide name:IL1B
- Sequence:118a.a.-269a.a.