Order Entry
Austria
ContactUsLinkComponent
Recombinant Human APOE2 (from E. coli)
Recombinant Human APOE2 (from E. coli)
Catalog # ABNOP7262.100UG
Supplier:  Abnova
CAS Number:  
Recombinant Human APOE2 (from E. coli)
Catalog # ABNOP7262.100UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Gene ID:
    NP_000032.1
  • Protein/peptide name:
    APOE2

Specifications

About this item