Order Entry
Austria
ContactUsLinkComponent
 
Recombinant Mouse TNFSF18 (from E. coli)
Catalog # ABNOP7358.10UG
Supplier:  Abnova
CAS Number:  
Recombinant Mouse TNFSF18 (from E. coli)
Catalog # ABNOP7358.10UG
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    10 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Gene ID:
    Q7TS55
  • Protein/peptide name:
    TNFSF18
  • Sequence:
    47a.a.-173a.a.

Specifications

About this item